missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2G12B Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-99414CL1
This item is not returnable.
View return policy
Description
PLA2G12B Polyclonal antibody specifically detects PLA2G12B in Mouse samples. It is validated for Western Blot
Specifications
| PLA2G12B | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| CoraFluor 1 | |
| group XIIB secretory phospholipase A2-like protein, group XIII secreted phospholipase A2, Group XIII secretory phospholipase A2-like protein, GXIIBMGC138151, GXIII sPLA2-like, GXIIIsPLA2, phospholipase A2, group XIIB, phospholipase A2, group XIII, PLA2G13, sPLA2-GXIIB | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Mouse PLA2G12B. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 84647 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction