missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2G16, Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse Polyclonal Antibody
Brand: Abnova H00011145-B01P.50ug
This item is not returnable.
View return policy
Description
Sequence: MRAPIPEPKPGDLIEIFRPFYRHWAIYVGDGYVVHLAPPSEVAGAGAASVMSALTDKAIVKKELLYDVAGSDKYQVNNKHDDKYSPLPCSKIIQRAEELVGQEVLYKLTSENCEHFVNELRYGVARSDQVRDVIIAASVAGMGLAAMSLIGVMFSRNKRQKQSpecifications
| PLA2G16 | |
| Polyclonal | |
| In 1x PBS, pH 7.4 | |
| BC001387 | |
| PLA2G16 | |
| HRASLS3 (AAH01387.1, 1 a.a. ∼ 162 a.a) full-length human protein. | |
| RUO | |
| 11145 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| Purified |
| Immunofluorescence, Western Blot | |
| Unconjugated | |
| phospholipase A2, group XVI | |
| AdPLA/H-REV107-1/HRASLS3/HREV107/HREV107-3/MGC118754 | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human | |
| Antibody |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction