missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2G1B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98077-50ul
This item is not returnable.
View return policy
Description
PLA2G1B Polyclonal antibody specifically detects PLA2G1B in Mouse samples. It is validated for ELISA
Specifications
| PLA2G1B | |
| Polyclonal | |
| ELISA 1:5000-1:10000 | |
| Group IB phospholipase A2, Phosphatidylcholine 2-acylhydrolase 1B, phospholipase A2, phospholipase A2, group IB (pancreas), PLA2AMGC119834, PLA2EC 3.1.1.4, PPLA2MGC119835 | |
| Produced in rabbits immunized with purified, recombinant Mouse PLA2G1B (Accession#: NP_035237.1; Met 1-Cys146) | |
| 50 μg | |
| Lipid and Metabolism | |
| 5319 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction