missing translation for 'onlineSavingsMsg'
Learn More

PLA2G2A Antibody, Novus Biologicals™

Goat Polyclonal Antibody

Brand:  Novus Biologicals NBP2-42399

Product Code. 18651199

  • ONLINE EXCLUSIVE PRICE
    € 531.00
    € 501.90 / 0.10mg
    Save € 29.10
    5% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PLA2G2A Polyclonal antibody specifically detects PLA2G2A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PLA2G2A
Polyclonal
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA
Goat
Immunogen affinity purified
RUO
Primary
Human
Purified
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide ELISA
0.5 mg/mL
Western Blot 0.1 - 0.3 μg/mL, Immunohistochemistry 5 μg/mL, Immunohistochemistry-Paraffin 5 μg/mL, Peptide ELISA Detection limit 1:8000
EC 3.1.1.4, GIIC sPLA2, Group IIA phospholipase A2, MOM1, Non-pancreatic secretory phospholipase A2, NPS-PLA2, Phosphatidylcholine 2-acylhydrolase 2A, phospholipase A2, group IIA (platelets, synovial fluid), phospholipase A2, membrane associated, PLA2BPLA2, PLA2LPLA2S, PLAS1, RASF-A, sPLA2
Peptide with sequence C-SYKFSNSGSRIT corresponding to internal region according to NP_000291.1.
0.1 mg
Lipid and Metabolism
5320
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.