missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PLA2G4A Monoclonal Antibody (C19)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA541580

 View more versions of this product

Product Code. 17867932

  • € 491.00 / 200µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PLA2G4A Monoclonal Antibody for Western Blot, IHC (P)

This gene encodes a member of the cytosolic phospholipase A2 group IV family. The enzyme catalyzes the hydrolysis of membrane phospholipids to release arachidonic acid which is subsequently metabolized into eicosanoids. Eicosanoids, including prostaglandins and leukotrienes, are lipid-based cellular hormones that regulate hemodynamics, inflammatory responses, and other intracellular pathways. The hydrolysis reaction also produces lysophospholipids that are converted into platelet-activating factor. The enzyme is activated by increased intracellular Ca(2+) levels and phosphorylation, resulting in its translocation from the cytosol and nucleus to perinuclear membrane vesicles.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PLA2G4A
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
P47712
PLA2G4A
Recombinant protein Cytosolic Phospholipase A2. The antigen corresponds to amino acid range 1-178 of the target protein.
200 μL
Primary
Human
Antibody
IgG2a κ
Immunohistochemistry (Paraffin), Western Blot
C19
Unconjugated
PLA2G4A
calcium-dependent phospholipid-binding protein; Cpla2; cPLA2alpha; cPLA2-alpha; cytosolic group IVA; Cytosolic phospholipase A2; group IVA; lysophospholipase; MGC126350; Phosphatidylcholine 2-acylhydrolase; Phospholipase A2; phospholipase A2 cytosolic; phospholipase A2 group IVA; phospholipase A2, group IVA (cytosolic, calcium-dependent); Pla2c; Pla2g4; PLA2G4A
Mouse
Protein A/G
RUO
5321
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.