missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2G4C Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93545-0.02ml
This item is not returnable.
View return policy
Description
PLA2G4C Polyclonal antibody specifically detects PLA2G4C in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| PLA2G4C | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| cPLA2-gamma, cytosolic phospholipase A2 gamma, DKFZp586C0423, EC 3.1.1.4, FLJ42247, FLJ44164, Phospholipase A2 group IVC, phospholipase A2, group IVC (cytosolic, calcium-independent) | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-189 of human PLA2G4C (NP_003697.2). MGSSEVSIIPGLQKEEKAAVERRRLHVLKALKKLRIEADEAPVVAVLGSGGGLRAHIACLGVLSEMKEQGLLDAVTYLAGVSGSTWAISSLYTNDGDMEALEADLKHRFTRQEWDLAKSLQKTIQAARSENYSLTDFWAYMVISKQTRELPESHLSNMKKPVEEGTLPYPIFAAIDNDLQPSWQEARAP | |
| 0.02 mL | |
| Endocrinology, Signal Transduction | |
| 8605 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction