Learn More
Invitrogen™ PLA2G4C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596790
Description
Immunogen sequence: MGSSEVSIIP GLQKEEKAAV ERRRLHVLKA LKKLRIEADE APVVAVLGSG GGLRAHIACL GVLSEMKEQG LLDAVTYLAG VSGSTWAISS LYTNDGDMEA LEADLKHRFT RQEWDLAKSL QKTIQAARSE NYSLTDFWAY MVISKQTREL PESHLSNMKK PVEEGTLPYP IFAAIDNDLQ PSWQEARAP; Positive Samples: THP-1; Cellular Location: Lipid-anchor, Membrane.
This gene encodes a protein which is a member of the phospholipase A2 enzyme family which hydrolyzes glycerophospholipids to produce free fatty acids and lysophospholipids, both of which serve as precursors in the production of signaling molecules. The encoded protein has been shown to be a calcium-independent and membrane bound enzyme. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Apr 2009].
Specifications
| PLA2G4C | |
| Polyclonal | |
| Unconjugated | |
| PLA2G4C | |
| cPLA2-gamma; Cytosolic phospholipase A2 gamma; DKFZp586C0423; FLJ42247; FLJ44164; phospholipase A2 group IVC; phospholipase A2, group IVC (cytosolic, calcium-independent); PLA2G4C | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 8605 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunocytochemistry | |
| 2.03 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UP65 | |
| PLA2G4C | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-189 of human PLA2G4C (NP_0036972). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.