missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2G4C Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539128
This item is not returnable.
View return policy
Description
This gene encodes a protein which is a member of the phospholipase A2 enzyme family which hydrolyzes glycerophospholipids to produce free fatty acids and lysophospholipids, both of which serve as precursors in the production of signaling molecules. The encoded protein has been shown to be a calcium-independent and membrane bound enzyme. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Apr 2009].
Specifications
| PLA2G4C | |
| Polyclonal | |
| Unconjugated | |
| PLA2G4C | |
| phospholipase A2 group IVC, cytosolic calcium-independent, DKFZp586C0423, FLJ42247, FLJ44164, cytosolic phospholipase A2 gamma, phospholipase A2 cytosolic calcium-independent gamma, phospholipase A2 group IVC | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8605 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q9UP65 | |
| PLA2G4C | |
| A synthetic peptide derived from the internal region of human PLA2G4C | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction