missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLA2G4D Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551226
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total PLA2G4D / cPLA2-delta protein.
The phospholipase A2 enzyme family, including PLA2G4D, catalyze the hydrolysis of glycerophospholipids at the sn-2 position and then liberate free fatty acids and lysophospholipids.[supplied by OMIM, Jun 2009].
Specifications
| PLA2G4D | |
| Polyclonal | |
| Unconjugated | |
| PLA2G4D | |
| 2610311B01Rik; cPLA2delta; cPLA2-delta; cytosolic phospholipase A2 delta; phospholipase A2 delta, cytosolic; phospholipase A2 group IVD; phospholipase A2, group IVD; phospholipase A2, group IVD (cytosolic); Pla2delta; PLA2G4D | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 283748 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.8 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q86XP0 | |
| PLA2G4D | |
| Fusion protein of human PLA2G4D. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction