missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLA2G6 Antibody (OTI10G4) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73456
This item is not returnable.
View return policy
Description
PLA2G6 Monoclonal specifically detects PLA2G6 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| PLA2G6 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1:2000 | |
| calcium-independent phospholipase A2, EC 3.1.1.4, Group VI phospholipase A2, IPLA2, iPLA285 kDa calcium-independent phospholipase A2, IPLA2-VIA, NBIA2A, NBIA2B, PARK14GVI, patatin-like phospholipase domain containing 9, phospholipase A2, group VI (cytosolic, calcium-independent), PNPLA9 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot | |
| OTI10G4 | |
| Unconjugated | |
| Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
| PLA2G6 | |
| Human recombinant protein fragment corresponding to amino acids 490-752 of human PLA2G6 (NP_001004426) produced in E.coli. | |
| 100 μg | |
| 8398 | |
| Human, Mouse, Rat | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction