missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PLA2G6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13482057
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13482057 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13482057 Supplier Invitrogen™ Supplier No. PA527945

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: U87-MG. Predicted reactivity: Mouse (94%), Rat (93%), Rabbit (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The protein encoded by this gene is an A2 phospholipase, a class of enzyme that catalyzes the release of fatty acids from phospholipids. The encoded protein may play a role in phospholipid remodelling, arachidonic acid release, leukotriene and prostaglandin synthesis, fas-mediated apoptosis, and transmembrane ion flux in glucose-stimulated B-cells. Several transcript variants encoding multiple isoforms have been described, but the full-length nature of only two of them have been determined to date.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLA2G6
Applications Western Blot
Classification Polyclonal
Concentration 0.74 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene PLA2G6
Gene Accession No. O60733
Gene Alias 85 kDa calcium-independent phospholipase A2; 85/88 kDa calcium-independent phospholipase A2; BB112799; CaI-PLA2; Group VI phospholipase A2; GVI; GVI PLA2; INAD1; intracellular membrane-associated calcium-independent phospholipase A2 beta; iPLA(2)beta; iPLA2; iPLA2beta; iPLA2-beta; IPLA2-VIA; NBIA2; NBIA2A; NBIA2B; neurodegeneration with brain iron accumulation 2; PARK14; patatin-like phospholipase domain-containing protein 9; phospholipase A2 group VI; phospholipase A2, group VI; phospholipase A2, group VI (cytosolic, calcium-independent); PLA2; PLA2G6; PLPLA9; PNPLA9
Gene Symbols PLA2G6
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PLA2G6. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8398
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.