missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLAA, Mouse anti-Human, Clone: 1F7, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00009373-M04.100ug
This item is not returnable.
View return policy
Description
Sequence: KNIHIALATLALNYSVCFHKDHNIEGKAQCLSLISTILEVVQDLEATFRLLVALGTLISDDSNAVQLAKSLGVDSQIKKYSSVSEPAKVSECCRFILNLLSpecifications
| PLAA | |
| Monoclonal | |
| Unconjugated | |
| phospholipase A2-activating protein | |
| DOA1/FLJ11281/FLJ12699/PLA2P/PLAP | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 1F7 | |
| In 1x PBS, pH 7.4 | |
| NM_004253 | |
| PLAA | |
| PLAA (NP_004244, 639 a.a. ∼ 738 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 9373 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction