missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLAC2 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76486AF405
This item is not returnable.
View return policy
Description
PLAC2 Polyclonal antibody specifically detects PLAC2 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| PLAC2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 405 | |
| FLJ90734, LINC00036, NCRNA00036, placenta-specific 2 (non-protein coding), PP10933 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the center of human PLAC2. The immunogen is located within amino acids 60 - 110 of PLAC2. Amino Acid Squence: WAARGHRKKRSTEGAGE | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 257000 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction