missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLAC4 Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41125AF750
This item is not returnable.
View return policy
Description
PLAC4 Polyclonal antibody specifically detects PLAC4 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| PLAC4 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 750 | |
| C21orf115, Chromosome 21 Open Reading Frame 115, D21S418E, Placenta-Specific 4, Placenta-Specific Protein 4, PRED78 | |
| Antibody was raised against a 17 amino acid synthetic peptide near the center of human PLAC4. The immunogen is located within amino acids 20 - 70 of PLAC4. Amino Acid Squence: DTLHPRHRHSSPWTPH | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 191585 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction