missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLAC8 Mouse anti-Human, Clone: OTI2E10, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810807S
This item is not returnable.
View return policy
Description
PLAC8 Monoclonal antibody specifically detects PLAC8 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| PLAC8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NZF1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 51316 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2E10 | |
| Unconjugated | |
| PLAC8 | |
| BM-004; C15; D5Wsu111e; DGIC; down-regulated in gastrointestinal cancer protein; onzin; PLAC8; placenta specific 8; placenta-specific 8; placenta-specific gene 8 protein; PNAS-144; Protein C15 | |
| Full length human recombinant protein of human PLAC8 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction