missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLAC8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567482
This item is not returnable.
View return policy
Description
Immunogen sequence: MNECCLCGTSV AMRTLYRTRY GIPGSICDDY MATLCCPHCT LCQIKRDINR RRAMRTF Highest antigen sequence identity to the following orthologs - mouse 84%, rat 86%.
PLAC8 is a protein coding gene.
Specifications
| PLAC8 | |
| Polyclonal | |
| Unconjugated | |
| PLAC8 | |
| BM-004; C15; D5Wsu111e; DGIC; down-regulated in gastrointestinal cancer protein; onzin; PLAC8; placenta specific 8; placenta-specific 8; placenta-specific gene 8 protein; PNAS-144; Protein C15 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51316 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NZF1 | |
| PLAC8 | |
| Recombinant Human PLAC8. Recombinant protein control fragment (Product #RP-108135). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction