missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PLAC9 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15915395
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15915395 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15915395 Supplier Invitrogen™ Supplier No. PA579825

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human placenta tissue, human placenta tissue.

By microarray assay to search for mouse genes enriched in placenta, Plac9 was identified as a placenta-enriched gene expressed in mouse 16-cell morula cDNA. By database analysis, human PLAC9, which encodes a deduced 97-amino acid protein, was identified. The deduced 108-amino acid mouse protein has a predicted signal peptide, a coiled-coil domain, and shares 73% identity with human PLAC9. Northern blot analysis of mouse placenta and embryo RNA at 12.5 and 18.5 days postcoitum (dpc) detected predominant expression in placenta.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLAC9
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene PLAC9
Gene Accession No. Q5JTB6
Gene Alias hypothetical protein LOC510496; PLAC9; Plac9a; placenta associated 9; placenta specific 9; placenta specific 9a; placenta-specific 9; placenta-specific protein 9
Gene Symbols PLAC9
Host Species Rabbit
Immunogen E. coli-derived human PLAC9 recombinant protein (Position: R30-L81).
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219348
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.