missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Placental Growth Factor Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10642-1-AP-20UL
This item is not returnable.
View return policy
Description
Placental growth factor (PlGF) is a member of the vascular endothelial growth factor (VEGF) family. PlGF is a pleiotropic factor affects different cell types and regulates various biological responses. The activity of vascular endothelial growth factor (VEGF), which is the most potent of all angiogenic mediators, is partly modulated by PlGF. It plays a major role in vasculogenesis and angiogenesis in human placenta.Specifications
| Placental Growth Factor | |
| Polyclonal | |
| Unconjugated | |
| PGF | |
| D12S1900, PGF, PGFL, PIGF, Placenta growth factor, placental growth factor, PLGF, PlGF 2, SHGC 10760 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5228 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P49763 | |
| PGF | |
| Placental Growth Factor Fusion Protein Ag0969 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction