Learn More
Tocris Bioscience™ Pladienolide B
Click to view available options
Quantity:
500 μg
Unit Size:
500µg
Description
Pladienolide B is a mRNA splicing inhibitor that decreases splicing capacity up to 75% in vitro. Pladienolide B directly targets splicesome-associated 130 (SAP130), inhibits splicing factor 3B subunit (SF3B1) and impairs U2 small nuclear ribonucleoprotein (U2 snRNP) interaction with pre-mRNA. Pladienolide B arrests the cell cycle in G1 and G2/M phases and displays antitumor activity against gastric cancer cells (IC50 values are 1.6-4.9 nM). Cell permeable and active in vivo.
Specifications
Specifications
| Chemical Name or Material | Pladienolide B |
| CAS | 445493-23-2 |
| Synonym | (4R,7R,8S,9E,11S,12S)-8-(Acetyloxy)-4,7-dihydroxy-12-[(1E,3E,5S)-6-[(2R,3R)-3-[(1R,2S)-2-hydroxy-1-methylbutyl]-2-oxiranyl]-1,5-dimethyl-1,3-hexadien-1-yl]-7,11-dimethyloxacyclododec-9-en-2-one |
| Quantity | 500 μg |
| Solubility | Soluble to 1mg/mL in DMSO |
| Molecular Formula | C30H48O8 |
| Formula Weight | 536.7 |
| Physical Form | Film |
| Color | Colorless |
| Product Type | Bioactive Small Molecule |
| Show More |