missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLAG1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584893
This item is not returnable.
View return policy
Description
Immunogen sequence: EPVDFLDPFT CNVSVPIKDE LLPVMSLPSS ELLSKPFTNT LQLNLYNTPF QSMQSSGSAH Q Highest antigen sequence indentity to the following orthologs: Rat - 97%, Mouse - 98%.
Pleomorphic adenoma gene 1 encodes a zinc finger protein with 2 putative nuclear localization signals. PLAG1, which is developmentally regulated, has been shown to be consistently rearranged in pleomorphic adenomas of the salivary glands. PLAG1 is activated by the reciprocal chromosomal translocations involving 8q12 in a subset of salivary gland pleomorphic adenomas. Three transcript variants encoding two different isoforms have been found for this gene.
Specifications
| PLAG1 | |
| Polyclonal | |
| Unconjugated | |
| PLAG1 | |
| C2H2 zinc finger protein PLAG1; COL1A2/PLAG1 fusion; HAS2/PLAG1 fusion; Plag1; PLAG1 zinc finger; pleiomorphic adenoma gene 1; pleiomorphic adenoma gene 1 protein; PSA; SGPA; Zinc finger protein PLAG1; ZNF912 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5324 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ChIP Assay, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6DJT9 | |
| PLAG1 | |
| Recombinant protein corresponding to Human PLAG1. Recombinant protein control fragment (Product #RP-107872). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion