Learn More
Fisherbrand™ Plain PTFE Stir Bars
Provides stable stirring action. Fisherbrand™ Plain PTFE Stir Bars provide greater turbulence than cylindrical stir bars.
Brand:  Fisherbrand™ 001.215.6
Product Code. 10068221
  • € 3.75 / Each
Only null left
Estimated Shipment: 27-01-2026
Description
  • Stable, general-purpose magnetic stir bar
  • Provides greater turbulence than cylindrical stir bars
  • Autoclavable