missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plakophilin-3 Antibody (12B11F8), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97675AF532
This item is not returnable.
View return policy
Description
Plakophilin-3 Monoclonal antibody specifically detects Plakophilin-3 in Human, Mouse, Porcine, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)
Specifications
| Plakophilin-3 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| plakophilin 3, plakophilin-3 | |
| This antibody is a mouse monoclonal IgG2a antibody derived by fusion of mouse myeloma cells with spleen cells from a mouse immunized with a synthetic peptide corresponding to the extreme carboxyterminal amino acid residues 779-793 (KLHRDFRAKGYRKED) of human plakophilin-3 coupled to keyhole limpet hemocyanin | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Pig, Canine | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Frozen) | |
| 12B11F8 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 11187 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction