missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plakophilin-3 Antibody (12B11F8), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97675R
This item is not returnable.
View return policy
Description
Plakophilin-3 Monoclonal specifically detects Plakophilin-3 in Human, Mouse, Porcine, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.
Specifications
| Plakophilin-3 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| PKP3 | |
| 12B11F8 is a mouse monoclonal IgG2a antibody derived by fusion of mouse myeloma cells with spleen cells from a mouse immunized with a synthetic peptide corresponding to the extreme carboxyterminal amino acid residues 779-793 (KLHRDFRAKGYRKED) of human pla | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Canine | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| 12B11F8 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen | |
| plakophilin 3, plakophilin-3 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 11187 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction