missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plasma Cell Antibody (LIV3G11), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50251CL1
This item is not returnable.
View return policy
Description
Plasma Cell Monoclonal antibody specifically detects Plasma Cell in Human samples. It is validated for Immunohistochemistry
Specifications
| Plasma Cell | |
| Monoclonal | |
| CoraFluor 1 | |
| Plasma | |
| Pancreatic cancer related serum mucin | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| Immunohistochemistry | |
| LIV3G11 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction