missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plasma Cell Antibody (LIV3G11), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50251UV
This item is not returnable.
View return policy
Description
Plasma Cell Monoclonal specifically detects Plasma Cell in Human samples. It is validated for Immunohistochemistry.
Specifications
| Plasma Cell | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Mouse | |
| Protein A purified | |
| Immunology | |
| Does not react with normal B cells. Reacts with normal and malignant human plasma cells | |
| Store at 4°C in the dark. |
| Immunohistochemistry | |
| LIV3G11 | |
| Immunohistochemistry | |
| Plasma | |
| Pancreatic cancer related serum mucin | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction