missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plasma Cell Marker, Mouse anti-Human, Clone: SPM310, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB15237.100ug
This item is not returnable.
View return policy
Specifications
| Plasma Cell Marker | |
| Monoclonal | |
| Unconjugated | |
| In 10 mM PBS. | |
| Pancreatic cancer-related mucin. | |
| RUO | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin) | |
| SPM310 | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.1-0.2 ug/mL) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction