missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plasma Cell Marker Mouse, Clone: LIV3G11; same as 7B18, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio MSM1-47-P1
This item is not returnable.
View return policy
Specifications
| Plasma Cell Marker | |
| Cytoplasmic | |
| LIV3G11; same as 7B18 | |
| Unconjugated | |
| Mouse | |
| Tonsil or lymph node | |
| 100 μg | |
| Primary | |
| IgG2a |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with 0.05% BSA and 0.05% azide | |
| Pancreatic cancer-related mucin | |
| Protein A/G Purified | |
| RUO | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction