missing translation for 'onlineSavingsMsg'
Learn More

Plasma Kallikrein/KLKB1 Antibody (JE31-29), Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-32506

Product Code. 30114631

  • € 519.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Plasma Kallikrein/KLKB1 Monoclonal antibody specifically detects Plasma Kallikrein/KLKB1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Plasma Kallikrein/KLKB1
Monoclonal
1 mg/mL
Western Blot 1:1000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200
EC 3.4.21, EC 3.4.21.34, Fletcher factor, kallikrein B, plasma (Fletcher factor) 1, kininogenin, KLK3plasma kallikrein, plasma kallikrein heavy chain, plasma kallikrein light chain, Plasma prekallikrein, PPK
Synthetic peptide within Human Plasma Kallikrein/KLKB1 aa 589-638/ 638. (Uniprot: P03952)
100 μL
Primary
Human
Purified
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
JE31-29
Unconjugated
1*TBS (pH7.4), 0.05% BSA and 40% Glycerol
Rabbit
Protein A purified
RUO
3818
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.