missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Plasminogen Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30044
This item is not returnable.
View return policy
Description
Plasminogen Polyclonal antibody specifically detects Plasminogen in Human samples. It is validated for ELISA,Western Blot
Specifications
| Plasminogen | |
| Polyclonal | |
| 75 mM Sodium Phosphate, 75 mM NaCl, 0.5 mM EDTA, pH 7.2 | |
| Goat | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| DKFZp779M0222, EC 3.4.21, EC 3.4.21.7, plasmin, plasminogen | |
| Human Plasminogen | |
| 1 mg | |
| Angiogenesis, Cancer, Cardiovascular Biology, Cell Cycle and Replication | |
| 5340 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction