missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Plasmolipin Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 188.00 - € 470.00
Specifications
| Antigen | Plasmolipin |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18622262
|
Novus Biologicals
NBP2-94002-0.02ml |
0.02 mL |
€ 188.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18692822
|
Novus Biologicals
NBP2-94002-0.1ml |
0.1 mL |
€ 470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Plasmolipin Polyclonal antibody specifically detects Plasmolipin in Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Plasmolipin | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS (pH 7.3), 50% glycerol | |
| 51090 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| Plasma membrane proteolipid, plasmolipin, PMLPTM4SF11plasma membrane proteolipid (plasmolipin), transmembrane 4 superfamily member 11 (plasmolipin) | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PLLP (NP_057077.1). MAEFPSKVSTRTSSPAQGAEASVSALRPDLGFVRSRLGALMLLQLVLGLLVWALIADTPYHLYPAYGWVMFVAVFLWLVTIVLFNLYLFQLHMKLYMVPW | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title