Learn More
Brady™ Plastic Mini Pictogram Signs
Product Code. p-8001198
Click to view available options
Height (Metric):
20 mm
50 mm
Unit Size:
Pack of 10
Pack of 15
Product Code. 10506710
Brand: Brady™ 802054
Only null left
Specifications
Certifications/Compliance BGV A 8 (VBG 125), DIN 4844
Alternate Part Number PIC 315-TRI 50-B-7586 10 ea/sheet
Material Plastic
Adhesive Type Permanent Acrylic
For Use With (Application) Used for pictograms and blank panels
Height (Metric) 50 mm
Thickness 0.026 mm
Width (Metric) 50 mm
Temperature Range (Metric) -20°C to 80°C
Shape Triangular
Displayed Text Warning signs
Product Type Mini Pictogram Safety Sign
Color Black on Yellow