missing translation for 'onlineSavingsMsg'
Learn More

TerraCycle™ Plastic Packaging Zero Waste Box™
GREENER_CHOICE

Product Code. 31935709
Change view
Click to view available options
Size:
Small
Medium
Large
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
31935709 Medium Each
31935700 Large Each
31935704 Small Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31935709 Supplier TerraCycle™ Supplier No. CS33MNL
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Support your sustainability goals and reduce landfill and incinerated waste with this easy-to-use recycling solution for Rigid Plastic Packaging and Soft Plastics. Zero Waste Box is an all-inclusive eco-friendly waste disposal service which includes storage, shipping and processing of the materials.

Use this Zero Waste Box to recycle:

  • Soft plastics
  • Rigid plastic packaging
  • Plastic bags
  • Bubble wrap

Do not send in: Biodegradable plastics, cardboard, paper, food content, liquid waste, metals, glass, plastic cards, label backing and any item contaminated with hazardous, flammable, or infectious substances.

Each box is supplied with a pre-affixed return shipping label.

Order: Select a Zero Waste Box™ and place an order.

Collect: Collect the accepted items to fill the box.

Ship: Once full, use the pre-paid shipping label (already fastened to the box) to arrange for your waste to be collected for recycling.

Reorder: Order your next Zero Waste Box™ to continue to recycle.

Everything collected is sorted and processed into raw materials that can be reused instead of being sent to landfill or incineration.

Cardboard boxes are made from 100% recycled material.

Specifications

Capacity (Metric) 78 L
Certifications/Compliance FSC
Capacity (English) 20.6 gal.
Color Brown, Green
Dimensions (L x W x H) 30.5 x 30.5 x 84 cm
Disposable Single-use
Latex Free Yes
Size Medium
Sterility Non-sterile
Autoclavable Not Autoclavable
Includes Return Shipping Label
For Use With (Application) Recycling Waste
Material Cardboard, Plastic
Product Type Recycling Box
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.