Learn More
Wuertt Celluloid™ Plasticized Drying Rack
Product Code. 10665221
Change view
Click to view available options
Height (Metric):
30 cm
61 cm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Height (Metric) Unit Size
10665221 30 cm Each
10734545 30 cm Each
10425591 61 cm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10665221 Supplier Wuertt Celluloid™ Supplier No. 52840111
Estimated Shipment: 20-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Benchtop rack and tray for drying laboratory glassware. Wuertt Celluloid™ Plasticized Drying Rack is constructed of plasticized wire.
  • Includes catch tray
  • Plasticized wire construction for durability
  • Angled stems
  • Looped stems for larger glassware
Specifications
Material Plasticized Wire
Positions 23
For Use With (Equipment) Laboratory glassware
No. per Pack 1 Pack
Quantity 1 Pack
Length (Metric) 42 cm
Depth (Metric) 17 cm
Includes Drain tray
Product Type Benchtop Drainer
Height (Metric) 30 cm