Learn More
Kartell™ Plastilab™ Untapered Y-Shaped Tubing Connectors
Description
Designed for use with hose system and vacuum lines.
- Made from polypropylene
- Autoclavable
- Nominal dimensions
Compliance
Meet Food and Agriculture regulations
Specifications
Specifications
| Certifications/Compliance | Meet with the Food and Drug regulations. 90/128/CEE 92/39/CEE e FDA 21 CFR 177.1520, CE and IVD |
| Autoclavable | Autoclavable |
| Material | Polypropylene |
| For Use With (Application) | Useful for any hoses system and vacuum line, suitable for food stuff |
| Packaging Quantity | 100/Pk. |
| Port Size | 2.7mm bore |
| Shape | Y |
| Product Type | Untapered Y Tubing Connector |