Learn More
BD Plastipak™ Concentric Luer-Slip Three-Piece Syringe
Product Code. p-8001027
Click to view available options
Volume (Metric):
2 mL
10 mL
Graduations:
0.1 mL
0.5 mL
Unit Size:
Pack of 100
Product Code. 10673555
Brand: BD 300185
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Use for sterile applications. BD Biosciences™ Plastipak™ Concentric Luer-Slip Three-Piece Syringe are individually wrapped and tested for sterility, pyrogenicity and toxicity.
  • Each syringe is tested for: sterility, pyrogenicity and toxicity
  • Syringes and separate accessory needles are individually wrapped in peel-apart blister packs.
  • For needles, please see our Microlance™ disposable needles range.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 20594-1, ISO 594-1 December 1993 ISO 594-2 September 1998
Material Polypropylene (Body)
Color Clear
Diameter (Metric) Outer 10 and 12 mm
Length (Metric) 60.15 mm
Product Type Concentric Luer-slip 3-piece Syringe
Disposable Single-use
Graduations 0.1 mL
Quantity 100/Pk.
Volume (Metric) 2 mL
Connection Type Concentric Luer Slip
Graduated Yes
Sterility Sterile