Learn More
PLAT Mouse anti-Human, Clone: OTI3H3, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF805448
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
PLAT gene encodes tissue-type plasminogen activator, a secreted serine protease that converts the proenzyme plasminogen to plasmin, a fibrinolytic enzyme. The encoded preproprotein is proteolytically processed by plasmin or trypsin to generate heavy and light chains. These chains associate via disulfide linkages to form the heterodimeric enzyme. This enzyme plays a role in cell migration and tissue remodeling. Increased enzymatic activity causes hyperfibrinolysis, which manifests as excessive bleeding, while decreased activity leads to hypofibrinolysis, which can result in thrombosis or embolism. Alternative splicing of this gene results in multiple transcript variants, at least one of which encodes an isoform that is proteolytically processed.Specifications
| PLAT | |
| Monoclonal | |
| Unconjugated | |
| PLAT | |
| T-PA, TPA | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5327 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3H3 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P00750 | |
| PLAT | |
| Human recombit protein fragment corresponding to amino acids 157-426 of human PLAT produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.