missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Plate Count Agar

Product Code. 17809682
Click to view available options
Quantity:
10/Pk.
Format:
90mm Monoplate
Media Type:
Plate Count Agar
Manufacturing Origin:
Germany
United Kingdom
Unit Size:
Pack of 10
This item is not returnable. View return policy

Product Code. 17809682

Brand: Thermo Scientific™ PO0158A

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Enumerate viable organisms in milk, water, food and dairy products using this non-selective medium.

Enumerate viable organisms in milk, dairy products, water, food and other specimens of sanitary importance using non-selective Thermo Scientific™ Plate Count Agar.

This standard medium meets the formulation of APHA, AOAC and PHLS.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Tested in accordance with CEN ISO/TS 11133-2.
Color Straw, clear
Content And Storage 2° to 10°C, away from light
Format 90mm Monoplate
Manufacturing Origin United Kingdom
Media Type Plate Count Agar
Product Type Agar
Fill Weight 19.5g ± 1.0g
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.