Learn More
Thermo Scientific™ Plate Frame for 96-well Piko PCR Plates, white
Shop All Thermo Scientific ProductsDescription
The Thermo Scientific Plate Frame for 96-well PikoPCR Plates, white, is designed for use with the Thermo Scientific Piko PCR Plates, a family of PCR plates which have the footprint of a microscope slide. We have not only shrunk the size of the plate, but dramatically reduced the thickness of the well walls utilizing our patented ultra-thin wall (UTW) technology.
Four plates can be snapped into a single frame producing the equivalent of a standard 384-well microplate.
Features
Designed for use with the Piko and PikoReal 96-well thermal cyclers
Plates can be snapped into the plate frame to create a standard 384-well plate
Compatible with standard multi-channel pipettes and liquid-handling platforms
Well spacing and footprint conform to industry (ANSI) dimensions
Applications
PCR
qPCR/Real-Time PCR
Related Products
Piko PCR Plate, 96-well, clear
Piko PCR Plate, 96-well, white
Four plates can be snapped into a single frame producing the equivalent of a standard 384-well microplate.
Features
Applications
Related Products
Piko PCR Plate, 96-well, clear
Piko PCR Plate, 96-well, white
Specifications
Specifications
| Packaging | 50 Frame |
| Product Line | Piko |
| Color | White |
| Product Type | Frame |
| For Use With (Equipment) | Piko Thermal Cycler, PikoReal Real-Time PCR System |
| Material | Polypropylene |
| No. of Wells | 96 |
| Plate Compatibility | 96-Well Plates |
| Quantity | 50 frames |