Learn More
Eppendorf™ Platforms for Innova™ 2300/43/43R, Excella™ E10/E25/E25R
Product Code. 11376004
Click to view available options
For Use With (Equipment):
4 L Erlenmeyer Flask Dedicated Platform
Unit Size:
Each
Product Code. 11376004
Brand: Eppendorf™ AG4
Eppendorf shaker platforms are available in a wide variety of materials and sizes, perfect for virtually every requirement. Universal platforms can be adapted for thousands of combinations of clamps and other accessories. Dedicated platforms come complete with a single clamp size pre-installed.
Platforms for Innova™ 2300/43/43R, Excella™ E10/E25/E25R, 76 × 46 cm, aluminum, 4 L Erlenmeyer flask dedicated platform- Flask Capacity: 6
TRUSTED_SUSTAINABILITY
Specifications
Material Aluminum
Dimensions (L x W) 76 x 46 cm
For Use With (Equipment) 4 L Erlenmeyer Flask Dedicated Platform