missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLAU Monoclonal Antibody (OTI6H1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805345S
This item is not returnable.
View return policy
Description
PLAU Monoclonal antibody specifically detects PLAU in Human samples. It is validated for Western BlotSpecifications
| PLAU | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P45381 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 443 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Western Blot | |
| OTI6H1 | |
| Unconjugated | |
| ASPA | |
| antagonist of uPA; ATF; BDPLT5; CD87; HGNC:9052; plasminogen activator; plasminogen activator, urinary; plasminogen activator, urokinase; Plau; QPD; uPA; u-PA; UPAM; UPAR; U-plasminogen activator; Urinary plasminogen activator, urokinase; URK; URKR; Urokinase-type plasminogen activator; Urokinase-type plasminogen activator chain B; Urokinase-type plasminogen activator long chain A; Urokinase-type plasminogen activator short chain A | |
| Human recombinant protein fragment corresponding to amino acids 77-313 of human ASPA produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction