missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLBD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571348
This item is not returnable.
View return policy
Description
PLBD1 Polyclonal Antibody for Western Blot
PLBD1 may act as an amidase or a peptidase that works with the small size of a putative binding pocket. PLBD1 exhibits a weak phospholipase activity, acting on various phospholipids, including phosphatidylcholine, phosphatidylinositol, phosphatidylethanolamine and lysophospholipids.
Specifications
| PLBD1 | |
| Polyclonal | |
| Unconjugated | |
| PLBD1 | |
| 1100001H23Rik; hypothetical protein; im:7138859; LAMA-like protein 1; Lamina ancestor homolog 1; phospholipase B domain containing 1; phospholipase B domain-containing protein 1; Phospholipase B-like 1; Phospholipase B-like 1 chain A; Phospholipase B-like 1 chain B; Phospholipase B-like 1 chain C; PLB homolog 1; PLBD1; putative phospholipase B-like 1; RGD1308734; zgc:194816 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 79887 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q6P4A8 | |
| PLBD1 | |
| synthetic peptide directed towards the C-terminal region of human PLBD1. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction