missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLC-beta 3 Antibody (SR1198), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22249-100ul
This item is not returnable.
View return policy
Description
PLC-beta 3 Monoclonal antibody specifically detects PLC-beta 3 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| PLC-beta 3 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| SR1198 | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
| 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-3, EC 3.1.4.11, FLJ37084, Phosphoinositide phospholipase C-beta-3, phospholipase C, beta 3 (phosphatidylinositol-specific), Phospholipase C-beta-3, PLC beta 3, PLC-beta-3 | |
| A synthesized peptide derived from human PLC-beta 3 (Uniprot #: Q01970) | |
| 100 μg | |
| Lipid and Metabolism, Phospho Specific | |
| 5331 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction