missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PLC-beta 4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 280.00
Specifications
| Antigen | PLC-beta 4 |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Regulatory Status | RUO |
Description
PLC-beta 4 Polyclonal specifically detects PLC-beta 4 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| PLC-beta 4 | |
| Unconjugated | |
| RUO | |
| Human | |
| 5332 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:SLPTIFCNIVLKTYVPDGFGDIVDALSDPKKFLSITEKRADQMRAMGIETSDIADVPSDTSKNDKKGKANTAKANVTPQSSSELRPTTTAALASGVEAKKGIELIPQVRIEDLKQMKAYLKHLKKQQKELNSLKKKHAKEHSTMQKL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism | |
| 1-phosphatidyl-D-myo-inositol-4,5-bisphosphate, 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta 4, 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-4, beta 4 (1-Phosphatidylinositol-4,5-BisphosphatePhosphodiesterase Beta 4)), EC 3.1.4.11, FLJ16169, inositoltrisphosphohydrolase, monophosphatidylinositol phosphodiesterase, phosphoinositidase C, phosphoinositide phospholipase C, phospholipase C, beta 4, triphosphoinositide phosphodiesterase | |
| PLCB4 | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title