missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLCB1 Polyclonal Antibody

Sheep Polyclonal Antibody
Brand: Invitrogen™ PA548052
This item is not returnable.
View return policy
Description
This antibody detects human and mouse PLC-beta 1, but does not detect recombinant human (rh) PLC-beta 2, rhPLC-beta 3, or rhPLC-beta 4 in Western blots. Reconstitute at 0.2 mg/mL in sterile PBS.
The protein encoded by this gene catalyzes the formation of inositol 1,4,5-trisphosphate and diacylglycerol from phosphatidylinositol 4,5-bisphosphate. This reaction uses calcium as a cofactor and plays an important role in the intracellular transduction of many extracellular signals. This gene is activated by two G-protein alpha subunits, alpha-q and alpha-11.
Specifications
| PLCB1 | |
| Polyclonal | |
| Unconjugated | |
| PLCB1 | |
| 1-phosphatidyl-D-myo-inositol-4,5-bisphosphate; 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase beta-1; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-1; 3110043I21Rik; AI132408; EIEE12; inositoltrisphosphohydrolase; KIAA0581; mKIAA0581; monophosphatidylinositol phosphodiesterase; Phosphb; phosphoinositidase C; phosphoinositide phospholipase C; phosphoinositide phospholipase C-beta-1; phosphoinositide-specific phospholipase c; phospholipase C; phospholipase C beta 1; phospholipase C, beta 1; phospholipase C, beta 1 (phosphoinositide-specific); Phospholipase C-beta1; phospholipase C-beta-1; phospholipase C-beta-1a; phospholipase C-I; PI-PLC; PLC'1; PLC154; PLC-154; Plcb; PLCB1; PLCB1A; PLCB1B; PLCbeta1; PLC-beta-1; PLC-I; RATPHOSPHB; triphosphoinositide phosphodiesterase | |
| Sheep | |
| Antigen affinity chromatography | |
| RUO | |
| 18795, 23236, 24654 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P10687, Q9NQ66, Q9Z1B3 | |
| PLCB1 | |
| E. coli-derived recombinant human PLC-beta 1 Lys27-Met245. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction