Learn More
PLCB2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27456-1-AP-20UL
 View more versions of this product
Product Code. 18084805
  • € 145.00 / 20µL
Description
The PLCB2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human PLCB2. This antibody recognizes human antigen. The PLCB2 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
Specifications
PLCB2
800 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, Immunofluorescence, ELISA
Phospholipase C beta 2, phospholipase C, beta 2, PLC beta 2, PLCB2
20 μL
5330
Store at -20°C. Stable for one year after shipment.