missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PLCB3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16360645
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16360645 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16360645 Supplier Invitrogen™ Supplier No. PA583691

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: RILVKNKKRH RPSAGGPDSA GRKRPLEQSN SALSESSAAT EPSSPQLGSP SSDSCPGLSN GEEVGLEKPS LEPQKSLGDE GLNRGPYVLG PADR Highest antigen sequence indentity to the following orthologs: Mouse - 84%, Rat - 88%.

PLCB3 encodes a member of the phosphoinositide phospholipase C beta enzyme family that catalyze the production of the secondary messengers diacylglycerol and inositol 1,4,5-triphosphate from phosphatidylinositol in G-protein-linked receptor-mediated signal transduction. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLCB3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Plcb3
Gene Accession No. Q01970
Gene Alias 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase beta-3; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase beta-3; EC 3.1.4.11; mKIAA4098; Phosphoinositide phospholipase C-beta-3; phosphoinositide-specific phospholipase c; phospholipase C beta 3; phospholipase C, beta 3; phospholipase C, beta 3 (phosphatidylinositol-specific); phospholipase C-beta-3; PLC beta 3; Plcb3; PLCbeta3; PLC-beta3; PLC-beta-3
Gene Symbols Plcb3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PLCB3. Recombinant protein control fragment (Product #RP-97385).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5331
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.