missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLCD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115769
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total PLCD1.
Phosphoinositide-specific phospholipase C (PLC) acts as a signal transducer that generates 2 second messengers, diacylglycerol and inositol 1, 4, 5-trisphosphate, by hydrolyzing inositol phospholipids. PLC comprises a diverse family of enzymes that differ in structure and tissue distribution (Berridge, 1993 ).
Specifications
| PLCD1 | |
| Polyclonal | |
| Unconjugated | |
| PLCD1 | |
| 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase delta-1; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase delta-1; AW212592; C79986; NDNC3; phosphoinositide phospholipase C; phosphoinositide phospholipase C-delta-1; phospholipase C delta 1; phospholipase C delta 1 Lf; phospholipase C delta 1 long form; phospholipase C, delta 1; Phospholipase C-delta1; phospholipase C-delta-1; phospholipase C-III; Plc1; Plcd; PLCD1; PLC-delta 1; PLC-delta-1; PLC-III | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18799, 24655, 5333 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P10688, P51178, Q8R3B1 | |
| PLCD1 | |
| A synthesized peptide derived from human PLCD1(Accession P51178), corresponding to amino acid residues H131-A181. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction