Learn More
Invitrogen™ PLCD4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110179
Description
Immunogen sequence: MASLLQDQLT TDQDLLLMQE GMPMRKVRSK SWKKLRYFRL QNDGMTVWHA RQARGSAKPS FSISDVETIR NGHDSELLRS LAEELPLEQG FTIVFHGRRS NLDLMANSVE EAQIWMRGLQ LLVDLVTSMD HQERLDQWLS DWFQRGDKNQ DGKMSFQEVQ RLLHLMNVEM DQEYAFSLFQ AADTSQSGTL EGEEFVQFYK ALTKRAEVQE LFESFSADGQ KLTLLEFLDF LQEEQKERDC TSELALELID RYEPSDSGKL RHVLSMDGFL SYLCSKDGDI FNPACLPIYQ.
This gene encodes a member of the delta class of phospholipase C enzymes. Phospholipase C enzymes play a critical role in many cellular processes by hydrolyzing phosphatidylinositol 4,5-bisphosphate into two intracellular second messengers, inositol 1,4,5-trisphosphate and diacylglycerol. Expression of this gene may be a marker for cancer.
Specifications
| PLCD4 | |
| Polyclonal | |
| Unconjugated | |
| Plcd4 | |
| 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase delta-4; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase delta-4; 4921507K24Rik; hPLCD4; phosphoinositide phospholipase C delta-4; phosphoinositide phospholipase C-delta-4; phospholipase C delta 4; phospholipase C, delta 4; phospholipase C-delta-4; PLC delta4; Plcd; PLCD4; PLC-delta-4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 140693, 18802, 84812 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q62711, Q8K3R3, Q9BRC7 | |
| Plcd4 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.