Learn More
Invitrogen™ PLCL2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140102
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: ITILKGQADL LKYAKNETLE NLKQIHFAAV SCGLNKPGTE NADVQKPRRS LEVIPEKAND ETGE Highest antigen sequence indentity to the following orthologs: Rat - 88%, Mouse - 89%.
May play an role in the regulation of Ins(1,4,5)P3 around the endoplasmic reticulum.
Specifications
| PLCL2 | |
| Polyclonal | |
| Unconjugated | |
| PLCL2 | |
| inactive phospholipase C-like protein 2; KIAA1092; mKIAA1092; phospholipase C; phospholipase C like 2; phospholipase C, epsilon 2; phospholipase C-epsilon-2; phospholipase C-L2; phospholipase C-like 2; PLCE2; PLC-epsilon-2; PLC-L(2); Plcl2; PLC-L2; PRIP-2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23228 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UPR0 | |
| PLCL2 | |
| Recombinant protein corresponding to Human PLCL2. Recombinant protein control fragment (Product #RP-109008). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.