missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PLCL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110180
This item is not returnable.
View return policy
Description
Immunogen sequence: TLDMLESSQD NMRTSWVSQM FSEIDVDNLG HITLCNAVQC IRNLNPGLKT SKIELKFKEL HKSKDKAGTE VTKEEFIEVF HELCTRPEIY FLLVQFSSNK EFLDTKDLMM FLEAEQGVAH INEEISLEII HKYEPSKEGQ EKGWLSIDGF TNYLMSPDCY IFDPEHKKVC Q.
May play an role in the regulation of Ins(1,4,5)P3 around the endoplasmic reticulum.
Specifications
| PLCL2 | |
| Polyclonal | |
| Unconjugated | |
| PLCL2 | |
| inactive phospholipase C-like protein 2; KIAA1092; mKIAA1092; phospholipase C; phospholipase C like 2; phospholipase C, epsilon 2; phospholipase C-epsilon-2; phospholipase C-L2; phospholipase C-like 2; PLCE2; PLC-epsilon-2; PLC-L(2); Plcl2; PLC-L2; PRIP-2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 224860 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.59 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q8K394 | |
| PLCL2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 130-300 of human PLCL2 (NP_055999.2). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction